missing translation for 'onlineSavingsMsg'
Learn More

Angelantoni Industrie™ VBH 48 C2 Compact Cabinet

Compact microbiological safety cabinet with a stainles steel work chamber and spillage tray. Angelantoni Industrie™ VBH 48 C2 Compact Cabinet features real-time, mircoprocessor-controlled parameters.

Marca:  Angelantoni Industrie™ ST-002900000

Encompass

Codice prodotto. 13555850

  • dietro preventivo / Pezzo

Effettua il per acquistare questo articolo Hai bisogno di un conto web ? Registrati oggi stesso!

Questo articolo richiede un aggiornamento del prezzo prima di essere ordinato.

Richiedi qui il prezzo per questo articolo.

Visualizzare prodotti alternativi »

Esplora altre offerte speciali
Questo articolo non è restituibile. Consulta la politica di reso

Descrizione

Descrizione

  • Fulfill and satisfy the European Norm EN12469 and country standards (presently superseded by EN12469) like German DIN 12950/10, French NF44-201 and English BS 5726
  • Manufactured with a frontal working opening for the aspiration of 30% of the total air involved, at a minimum velocity of 0,40 m/s, with the remaining 70% recirculated through a HEPA filter in the working chamber
  • Mean laminar flow velocity of 0,40 m/s (as defined by European standard EN 12469: 2000)
  • Containment index, evaluated on the total surface of the front working opening, corresponds to APF (Aperture Protection Factor) equal or more than 1 x 105 according EN12469
  • Microprocessor monitors all critical parameters of the unit (laminar air flow velocity, exhaust flow rate and position of the front screen) in real time and controls the optimal function of the fans even if in presence of effects caused by progressive clogging of the HEPA filters
  • Work chamber and spillage tray in stainless steel AISI 304L 2B finishing
  • Working surface is removable and autoclavable
  • Hinged front window, in laminated safety glass; ergonomic opening 150°
  • Maximum free height with open screen: 540 mm
  • Full vision height of 535 mm at operational condition
  • Ergonomic front closure panel in anodized aluminum complete of germicidal UV lamp
  • Lighting on working surface by means of 2 fluorescent electronic lamps fitted in non-contaminated area
  • Construction structure in epoxy-polyester powder painted steel
  • Ventilation system by means of n°2 IP55 protection factor motor blowers
  • Recirculating and exhaust air filtration achieved by H14 HEPA filters as per EN 1822 (with efficiency 99.995% MPPS)
  • Maintenance completely from the front of the cabinet
Specifica

Specifica

EN 12469, DIN 12950/10, NF 44-201, BS 5726, EN 1822
890 mm
VBH 48 C2
HEPA
UV Lamp
900 mm
760 mm
685 mm
Stainless Steel
Safety Cabinet
Double HEPA
manipulation of QC in pharmaceutical and food industries, micro mechanical assembly, and DNA/RNA amplification and thermo-cyclling
156.9 cm
760 x 1050 x 1590 mm
1050 mm
Suggerimenti di prodotto

Suggerimenti di prodotto

Videos
SDS
Documenti

Documenti

Certificati
Offerte speciali

Offerte speciali

Correzione del contenuto del prodotto

Il programma Encompass di Fisher Scientific offre articoli che non fanno parte del nostro portafoglio di distribuzione. Questi prodotti in genere non hanno immagini o descrizioni dettagliate. Tuttavia, ci impegniamo a migliorare la vostra esperienza di acquisto. Si prega di utilizzare il modulo sottostante per fornire un feedback relativo al contenuto di questo prodotto.

Titolo del prodotto
Angelantoni Industrie™ VBH 48 C2 Compact Cabinet

Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.

Grazie per averci aiutato a migliorare il nostro sito web. Il vostro feedback è stato inviato