Learn More
DWK Life Sciences Watch Glass Dish, made from soda-lime glass, fused rim
The concave shape of the watch glass dish allow working in very small quantities (e.g. for pretreatments) and covering laboratory glassware with wide openings.
Marca:  DWK Life Sciences 233214307
28.88 EUR valido fino al 2024-12-31
Usa il codice promo "23258" per avere il tuo prezzo promozionale.
Dettagli aggiuntivi : Peso : 0.30000kg
Codice prodotto. 15211849
  • 30.40€ / pezzi 10
Spedizione stimata: 21-08-2024
per visualizzare lo stock disponibile
Aggiungi al carrello
Descrizione
Descrizione
The multitude of different work steps in the laboratory environment demands a multitude of universally applicable glassware. These glass devices usually have a very simple shape and are characterized by their versatility. Watch glasses, for example, are concave curved. The small trough created in this way allows liquids or solids to be filled in.
  • Reduced risk of injury due to fused rim
Specifica
Specifica
Not Autoclavable
Weighing; Covering; Pretreatments; Drying of materials; Available in Boro 3.3 or soda-lime glass
90 mm
Soda Lime Glass
10/Pk.