Learn More
Cytiva Whatman™ AS 600 Series Filtration Manifold
Stainless steel filter funnel six-place manifold, for microbiology monitors and analytical funnels
Marca: Cytiva 10498762
1921.13 EUR valido fino al 2025-04-30
Usa il codice promo "24138" per avere il tuo prezzo promozionale.
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
The stainless steel manifold for three or six filtration units is fitted with stainless steel units. The apparatus can be autoclaved and sterilized by dry heat at up to 180°C. Suitable only for vacuum operation. If flushing tubes are used, do not exceed 1.3 bar (300 mbar over-pressure).
Specifica
Autoclavable | |
Stainless Steel | |
9 mm I.D. |
Microbiology monitors and analytical funnels | |
6 Stopcocks and lower parts for individual choice of filter units |
Titolo del prodotto
Cytiva Whatman™ AS 600 Series Filtration Manifold > Six-place filtration manifold