Learn More
ABEBA™ X-Light Black/Yellow Safety Trainers S3
Black/yellow X-light safety trainer with silver ion lining to reduce odor, replaceable active comfort insole, steel toe cap, textile antipuncture midsole, PU slip resistant outsole
Marque: ABEBA™ 711860/47
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
- Lightweight, comfortable trainers
- Comply to EN ISO 20345: 2012, S3 SRC
- Slip resistant Mono PU outsole SRC
- Steel toe cap (200 J)
- Textile antipuncture midsole
- Black and yellow Cordura
- Silver ions lining
- Replaceable active comfort insole
Spécification
Black, Yellow | |
47 | |
Comply to EN 20345: 2012 S3 SRC | |
Trainers | |
S3 SRC |
Cordura | |
Steel | |
Unisex | |
Water Resistant |