CAS RN 1044589-82-3
CAS RN 1044589-82-3
IUPAC Name:
—
Sinonimi:
—
Peso molecolare (g/mol):
—
Formula molecolare:
—
InChi Key:
—
SMILES:
—
1
–
3
di
3
risultati
1
Tocris Bioscience™ NITD008
Antiviral; selective flavivirus inhibitor
2
NITD008, MedChemExpress
MedChemExpress NITD008 is a potent and selective flaviviruse inhibitor which can inhibit Dengue Virus Type 2 (DENV-2) with an EC50 of 0.64 μM.
3