missing translation for 'onlineSavingsMsg'
Learn More

2-Phenylethyl isothiocyanate, 98%, Thermo Scientific Chemicals

49.95€ - 563.00€

Identificatori chimici

CAS 2257-09-2
Formula molecolare C9H9NS
Molecular Weight (g/mol) 163.24
Numero MDL MFCD00004821
InChI Key IZJDOKYDEWTZSO-UHFFFAOYSA-N
Sinonimo 2-phenylethyl isothiocyanate, phenethyl isothiocyanate, phenylethyl isothiocyanate, 2-isothiocyanatoethyl benzene, phenethyl mustard oil, phenylaethylsenfoel, benzene, 2-isothiocyanatoethyl, peitc, phenylethyl mustard oil, 2-phenylethylisothiocyanate
PubChem CID 16741
ChEBI CHEBI:351346
IUPAC Name 2-isothiocyanatoethylbenzene
SMILES C1=CC=C(C=C1)CCN=C=S
Visualizza altre specifiche

Prodotti 3
Codice del prodotto Marca Quantità Prezzo Quantità e disponibilità  
Codice del prodotto Marca Quantità Prezzo Quantità e disponibilità  
11422088
Vedere i documenti
Thermo Scientific Alfa Aesar
A15123.06
5 g
49.95€
5g
Effettua il per acquistare questo articolo Hai bisogno di un conto web ? Registrati oggi stesso!
11432088
Vedere i documenti
Thermo Scientific Alfa Aesar
A15123.14
25 g
191.96€
25g
Effettua il per acquistare questo articolo Hai bisogno di un conto web ? Registrati oggi stesso!
11442088
Vedere i documenti
Thermo Scientific Alfa Aesar
A15123.22
100 g Questo articolo non è attualmente disponibile o è stato dismesso.
La preghiamo di consultare la pagina del prodotto per individuare le possibili alternative.
N/A
Descrizione

Descrizione

Phenethyl isothiocyanate is an NNK-induced lung tumorigenesis inhibitor.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Phenethyl isothiocyanate is an NNK-induced lung tumorigenesis inhibitor.

Solubility
Soluble in chloroform, ethyl acetate, and dichoromethane. Reacts with water.

Notes
Moisture Sensitive, store away from moisture. Store in cool, dry place in tightly closed container. Store away from oxidizing agent. Store under dry inert gas.
Specifica

Identificatori chimici

2257-09-2
163.24
IZJDOKYDEWTZSO-UHFFFAOYSA-N
16741
2-isothiocyanatoethylbenzene
C9H9NS
MFCD00004821
2-phenylethyl isothiocyanate, phenethyl isothiocyanate, phenylethyl isothiocyanate, 2-isothiocyanatoethyl benzene, phenethyl mustard oil, phenylaethylsenfoel, benzene, 2-isothiocyanatoethyl, peitc, phenylethyl mustard oil, 2-phenylethylisothiocyanate
CHEBI:351346
C1=CC=C(C=C1)CCN=C=S

Specifica

2-Phenylethyl isothiocyanate
139°C to 140°C (11 mmHg)
Pungent
MFCD00004821
2084162
IZJDOKYDEWTZSO-UHFFFAOYSA-N
2-isothiocyanatoethylbenzene
16741
163.24
2257-09-2
>110°C (230°F)
C9H9NS
UN2922
2-phenylethyl isothiocyanate, phenethyl isothiocyanate, phenylethyl isothiocyanate, 2-isothiocyanatoethyl benzene, phenethyl mustard oil, phenylaethylsenfoel, benzene, 2-isothiocyanatoethyl, peitc, phenylethyl mustard oil, 2-phenylethylisothiocyanate
C1=CC=C(C=C1)CCN=C=S
163.24
CHEBI:351346
98%
Videos
Sicurezza e movimentazione

Sicurezza e movimentazione

GHS H Statement
H331-H314-H318-H302-H312
Toxic if inhaled.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
Harmful in contact with skin.

GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

missing translation for 'dotInformation' : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE LIQUIDS, TOXIC, N.O.S.

missing translation for 'rtecsNumber' : NX9115000

missing translation for 'tsca' : TSCA

Avvertenza
  • Pericolo

Categoria di pericolo
  • Tossicità acuta Categoria 4
  • Lesione oculare grave/irritazione oculare Categoria 2
  • Sensibilizzante delle vie respiratorie Categoria 1
  • Corrosione/irritazione della pelle Categoria 2
  • Tossicità specifica per organi bersaglio (esposizione singola) Categoria 3

Indicazioni di pericolo
  • H302-Nocivo se ingerito.
  • H312-Nocivo per contatto con la pelle.
  • H315-Provoca irritazione cutanea.
  • H319-Provoca grave irritazione oculare.
  • H332-Nocivo se inalato.
  • H334-Può provocare sintomi allergici o asmatici o difficoltà respiratorie se inalato.
  • H335-Può irritare le vie respiratorie.

Consigli di prudenza
  • P280-Indossare guanti/indumenti protettivi/Proteggere gli occhi/il viso.
  • P284-¢Quando la ventilazione del locale è insufficiente! indossare un apparecchio di protezione respiratoria.
  • P301+P330+P331-IN CASO DI INGESTIONE: sciacquare la bocca. NON provocare il vomito.
  • P302+P352-IN CASO DI CONTATTO CON LA PELLE: lavare abbondantemente con acqua/.
  • P304+P340-IN CASO DI INALAZIONE: trasportare l'infortunato all'aria aperta e mantenerlo a riposo in posizione che favorisca la respirazione.
  • P305+P351+P338-IN CASO DI CONTATTO CON GLI OCCHI: sciacquare accuratamente per parecchi minuti. Togliere le eventuali lenti a contatto se è agevole farlo. Continuare a sciacquare.
  • P312-Contattare un CENTRO ANTIVELENI/un medico/in caso di malessere.

SDS
Documenti

Documenti

Product Certifications

RUO – Research Use Only

Correzione del contenuto del prodotto

Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.

Titolo del prodotto

Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.

Grazie per averci aiutato a migliorare il nostro sito web. Il vostro feedback è stato inviato