missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Methyl-1-Butanol, 98%, Reagent Grade, Honeywell™
Reagent Grade, 98%
22.38€ - 211.09€
Identificatori chimici
CAS | 123-51-3 |
---|---|
Formula molecolare | C5H12O |
Molecular Weight (g/mol) | 88.15 |
Numero MDL | MFCD00002934 |
InChI Key | PHTQWCKDNZKARW-UHFFFAOYSA-N |
Sinonimo | 3-methyl-1-butanol, isoamyl alcohol, isopentyl alcohol, isopentanol, 3-methylbutanol, 1-butanol, 3-methyl, isoamylol, isobutylcarbinol, iso-amylalkohol, isobutyl carbinol |
PubChem CID | 31260 |
ChEBI | CHEBI:15837 |
IUPAC Name | 3-methylbutan-1-ol |
SMILES | CC(C)CCO |
Codice del prodotto | Marca | Quantità | Imballaggio | Prezzo | Quantità e disponibilità | ||||
---|---|---|---|---|---|---|---|---|---|
Codice del prodotto | Marca | Quantità | Imballaggio | Prezzo | Quantità e disponibilità | ||||
15677940
|
Honeywell Research Chemicals
M32658-100ML |
100 mL | Glass bottle | N/A | |||||
15607950
|
Honeywell Research Chemicals
M32658-500ML |
500 mL | Glass bottle | N/A | |||||
15687940
|
Honeywell Research Chemicals
M32658-1L |
1 L | Glass bottle | N/A | |||||
15697940
|
Honeywell Research Chemicals
M32658-2.5L |
2.5 L | Glass bottle | N/A | |||||
Descrizione
- ≥95% for general laboratory use.
Identificatori chimici
123-51-3 | |
88.15 | |
PHTQWCKDNZKARW-UHFFFAOYSA-N | |
31260 | |
3-methylbutan-1-ol |
C5H12O | |
MFCD00002934 | |
3-methyl-1-butanol, isoamyl alcohol, isopentyl alcohol, isopentanol, 3-methylbutanol, 1-butanol, 3-methyl, isoamylol, isobutylcarbinol, iso-amylalkohol, isobutyl carbinol | |
CHEBI:15837 | |
CC(C)CCO |
Specifica
3-Methyl-1-butanol | |
-117°C | |
0.809g/mL at 25°C | |
1.2 to 9% (100°F) | |
644°F | |
(CH3)2CHCH2CH2OH | |
UN1105 | |
3-methyl-1-butanol, isoamyl alcohol, isopentyl alcohol, isopentanol, 3-methylbutanol, 1-butanol, 3-methyl, isoamylol, isobutylcarbinol, iso-amylalkohol, isobutyl carbinol | |
CC(C)CCO | |
88.15 | |
3 (vs air) | |
88.15g/mol | |
0.98 |
123-51-3 | |
Colorless | |
131°C to 132°C | |
43°C (109.4°F) | |
C5H12O | |
MFCD00002934 | |
1718835 | |
PHTQWCKDNZKARW-UHFFFAOYSA-N | |
3-methylbutan-1-ol | |
31260 | |
CHEBI:15837 | |
2mmHg (20°C) |
Sicurezza e movimentazione
P210-P261-P280-P304 + P340 + P312-P337 + P313-P403 + P235
missing translation for 'dotInformation' : class 3 - PG 3 - Pentanols
missing translation for 'rtecsNumber' : 204-633-5