Learn More
Thermo Scientific™ Accessories for Orion™ pH Electrodes
Store and protect your pH electrodes with these accessories.
Marca: Thermo Scientific™ 910005
41.82 EUR valido fino al 2025-04-30
Usa il codice promo "24138" per avere il tuo prezzo promozionale.
Descrizione
Store and protect your pH electrodes with the Thermo Scientific™ Accessories for Orion™ pH Electrodes. Storage bottles protect and condition electrodes when not in use. Bulb guard protects the pH-sensing bulb of glass pH electrodes.
Electrode Bulb Guards
- Designed for glass body pH electrodes
- Use with standard or Thermo Scientific™ Orion™ Sure-Flow™ pH electrodes — one end of the bulb guard fits over standard electrodes while the other end fits over Sure-Flow electrodes without interfering with the Sure-Flow flushing action
Specifica
Bulb guard for glass pH Electrodes | |
Sure-Flow and Standard Glass Electrodes | |
Bulb Guard |
Offerte speciali
-
Diversi produttori
Back to the Lab Summer Semester
Save up to 94% on Teaching Lab EssentialsCodice sconto: 25721Scade: 2025-05-01
nuova offerta Save Now