Learn More
Memmert™ Additional Stainless Steel Grid for UN260, Reinforced Chamber Option
Additional grid features a sturdy stainless steel construction.
Marca: Memmert™ UN260E29766
Descrizione
- Reinforced
- Stainless Steel
- Electropoloished
- Max. loading 60 kg
Specifica
UN260 Oven | |
Reinforced Grid | |
Acciaio inox | |
60 kg |