Learn More
Abeba™ Arrow 38210 Shoes
Antistatic with heel patch microfiber. Sanitized™ treated.
Marca: Abeba™ 38210/40
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
- CE EN ISO 20347:2012 OB A, FO SRA
- Smooth leather
- Black
- Slip resistance PU/PU outsole sole
- Sanitized treated
- ESD dissipative according to DIN EN 61340
- Heel strap foldable, adjustable
- Microfiber heel patch
- Lining microfiber, respiration absorbing
Specifica
CE EN ISO 20347:2012 OB A, FO SRA | |
Microfiber | |
40 | |
Polyurethane |
Black | |
Leather | |
Slip-resistant | |
38210 |
Suggerimenti di prodotto
Customers who viewed this item also viewed.
Viewing 1-4 of
Titolo del prodotto
Abeba™ Arrow 38210 Shoes > Size: 40