missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IBA Lifesciences CD4 Fab Streptamer™ Isolation Kit MB
CD4 Fab Streptamer Isolation Kit MB is for reversible cell isolation delivering label-free target cells.
Marca: IBA Lifesciences 6-8000-206
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
- Strep-Tactin™ Magnetic Microbeads for Fab Streptamers™; 2 vials for 5 x 108 cells each
- CD4 Fab-Strep, human for isolation of T-cells; 1 vial for 1 x 109 cells
- Biotin stock solution for Streptamer™ technology; 2 x 1 mL
- Buffer IS, Streptamer™ Washing Buffer, 10x concentrated; 50 mL
- Microbeads are intended for use with a permanent magnet for manual cell isolation; we recommend our StrepMan magnet for 2 x 15 mL and 2 x 50 mL Falcon tubes.
- Principle of Selection: Isolation of human CD4 T cells is performed by positive selection.
- PBMCs are labeled with magnetic CD4 Fab Streptamers™.
- Labeled cells are isolated in a strong magnet (StrepMan, horizontal position) where they migrate toward the tube wall on the side of the magnet.
- This CD4 positive cell fraction is collected and cells are liberated from all labeling reagents by addition of biotin in a strong magnet (StrepMan, upright position).
- The magnetic Streptamers™ migrate toward the tube wall and the label-free cells remain in the supernatant.
- Biotin is removed by washing.
- The resulting cell preparation is highly enriched with CD4+ T cells with a purity of more than 90%.
- No depletion steps and no columns are needed.
Specifica
PBMCs | |
Magnetic Microspheres Coated with Strep-Tactin | |
1 x 109 | |
1 Kit |
Suggerimenti di prodotto
I clienti che hanno visualizzato questo articolo hanno visto anche
Manual PR32 Fab Streptamer Microbeads 6-8000-206
Dimensione file: 1.2 MB
Short Protocol PR40-FabStreptamerMicrobeads 6-8000-206
Dimensione file: 1.2 MB
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto
IBA Lifesciences CD4 Fab Streptamer™ Isolation Kit MB > Cell Isolation Kit
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto