missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Proteintech CoraLite™594 TUNEL Assay Apoptosis Detection Kit
A remarkable feature of apoptosis is the degradation of chromosome DNA, which is a common phenomenon.
Marca: Proteintech PF00009-50T
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
- The degradation is very specific and regular. DNA fragments break into different lengths are of 180 bp-200 bp, showing a specific ladder like ladder pattern in agarose gel electrophoresis.
- The kit uses TUNEL method and uses Terminal Deoxynucleotidyl Transferase(TdT) catalyzes the incorporation of CoraLite™594-dUTP into the 3'-OH terminal of the broken DNA of apoptotic cells.
- DNA labeled with CoraLite™594- dUTP can be observed directly by fluorescence microscope or quantified by flow cytometry.
- TUNEL can selectively detect apoptotic cells rather than necrotic cells or cells with DNA strand breaks caused by irradiation and drug therapy.
Specifica
Emission: 614 nm | |
50 Tests | |
Flow Cytometry | |
Store at -20°C. Avoid exposure to light. Stable for 2 years after shipment. Avoid freeze- thaw cycles. |
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto
Proteintech CoraLite™594 TUNEL Assay Apoptosis Detection Kit > 50 Tests
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto