Learn More
Greiner Bio-One Plasma Tubes with Lithium Heparin and Gel
For determinations in clinical chemistry and immunochemistry
Marca:  Greiner Bio-One 455083
Codice prodotto. 15433180
  • 38.00€ / pezzi 50
Esplora altre offerte speciali
Questo articolo non è restituibile. Consulta la politica di reso
Descrizione
Descrizione
  • Interior tube wall is coated with lithium heparin, an anticoagulant that binds antithrombins to block the coagulation cascade producing a plasma sample
  • Contain a barrier gel in the tube that moves upward to form a stable barrier separating plasma from cells during centrifugation; plasma may be aspirated directly from the tube eliminating the need for transfer
  • Clear as glass with equal or better performance
  • Molded to be virtually unbreakable, providing a better level of safety than glass
  • Tubes have an 18-month shelf life from the date of manufacture
  • Nonscrew Safety Pull Caps can be removed with a gentle pull
TRUSTED_SUSTAINABILITY
Specifica
Specifica
Lithium Heparin
8 mL
16 x 100 mm
Safety Pull Cap
Green with Yellow Ring
16 x 100 mm
Titolo del prodotto
Greiner Bio-One Plasma Tubes with Lithium Heparin and Gel > Fill Volume: 8mL; Pull closure; Size: 16 x 100mm