missing translation for 'onlineSavingsMsg'
Learn More
Learn More
WTW™ inoLab™ 7110 Benchtop pH Meter

A simple, easy-to-use lab pH meter for routine measurement with reproducible measuring results and increased measuring accuracy.
Marca: WTW™ 1AA114
1066.00 EUR valido fino al 2025-12-31
Usa il codice promo "25339" per avere il tuo prezzo promozionale.
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
- Repeatable measuring results by automatic recognition of stable values
- Secure operation: automated functions reduce the number of keys
- Increased measuring accuracy by calibration timer with periodic reminders for calibration
- Highly suitable for routine measurements in the lab, where automatic documentation is not a priority
- With smooth, easy to clean surface
- Available as a single meter, as a set with standard electrode with built-in T sensor, or as a set with a precision electrode

Specifica
DIN socket, Universal power supply, electrode arm and operation manual, universal power supply, electrode arm and operation manual, 3 in 1 pH electrode SenTix™ 81, buffers, 3mol/L KCl, cable | |
±0.3, ±1 | |
Current record calibration memory | |
-2 to 20 | |
±1200, ±0.3 | |
Da 1 a 3 | |
CE, cETLUS | |
3 Years | |
Automatic | |
DIN or BNC/4mm banana | |
240 mm | |
80 mm |
±0.1K | |
±0.005, ±0.01 | |
Ideal for routine measurements in the laboratory where automatic documentation is not the priority and for AC and battery operation | |
-2 to 20 ±0.01, -2 to 19.999 ±0.005 | |
±(1200.0 ±0.3mV) | |
LCD | |
1 kg | |
AC power adapter or four AA batteries | |
-5°C to +105°C | |
Benchtop pH meter | |
190 mm |
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto
WTW™ inoLab™ 7110 Benchtop pH Meter > Calibration Points: 1, 2 or 3; Power Supply: AC power adapter or four AA batteries; Includes: 3 in 1 pH electrode SenTix 81, buffers, 3mol/L KCl, cable
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto