Learn More
DWK Life Sciences Kimble™ KIMAX™ Pycnometer Bottle
Designed for measuring and comparing the densities or specific gravities of liquids and solids. This non-mercury spirit-filled pycnometer is supplied with Standard Taper 10/18 thermometer with a range of 14 to 38°C in 0.2° increments.
Marca: DWK Life Sciences 15123N-25
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
- Thermometer is retested for accuracy within ±0.2 degrees C
- Standard Taper 5/12 side tube cap has a small vent near the top
- Body is conical for maximum stability
- Capacity is within 10 percent of nominal shown on the flask
- Each flask body is numbered for ready identification
Specifica
25 mL | |
175 mm | |
Pycnometer Bottle |
Titolo del prodotto
DWK Life Sciences Kimble™ KIMAX™ Pycnometer Bottle > Capacity: 25mL
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto