Learn More
Novus Biologicals™ DGCR8 Knockout MEF Cells
Applications: In vitro assay
Marca: Novus Biologicals™ NBP2-25171
Descrizione
- Cells are supplied in 1.5mL quantities (about 1 x 106 cells/mL) in freezing media (80% MEF media, 20% FBS, 20% DMSO)
- Gene ID: 54487
- Gene symbol: DGCR8
Recommended Storage
Store in gas phase of liquid nitrogen
Servizi
6 Months

Specifica
Store in gas phase of liquid nitrogen | |
In vitro assay | |
DGCR8 Knockout MEF Cells | |
1.5mL |
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
This product is for research use only and is not approved for use in humans or in clinical diagnosis