Learn More
Kolmi™ Op Air Pro Oxygen FFP2 NR D Mass Balance Masks
Op Air Pro Oxygen FFP2 masks are easy to put on with high breathability due to high quality raw material. Made with the Mass Balance approach from recycled materials.
Marca: Kolmi™ M52114S-WH-MB
19.67 EUR valido fino al 2025-06-08
Usa il codice promo "24131" per avere il tuo prezzo promozionale.
Descrizione
- Filtering half mask, efficiency class FFP2 NR D developed to protect against solid and liquid aerosols and particles
- Type IIR certified, it protects the environment against droplets emitted by the wearer and it protects from potential body fluid splashes
- Knitted headloops ultrasonically welded for optimal comfort
- Fits perfectly to face to limit inward leakage thanks to its shape and soft materials
- Hypoallergenic and has been dermatologically tested
- Packaged in individual polybags
- Made in France
Specifica
EN 149: 2001 + A1: 2009, DM Class I, EN 14683: 2019 + AC: 2019 | |
Large | |
Headband |
White | |
Pouch |
Offerte speciali
-
Diversi produttori
Gear Up for Safety
Save up to 60% on gloves, apparel, and moreCodice sconto: 24131Scade: 2025-06-08
nuova offerta Shop Now