Learn More
Fisherbrand™ Siringhe in PP in plastica, Luer Lock
Siringhe monouso non sterili con certificazione HPLC realizzate in polipropilene resistente ai solventi
Marca: Fisherbrand™ 02 36 2399
25.11 EUR valido fino al 2025-05-31
Usa il codice promo "24127" per avere il tuo prezzo promozionale.
Descrizione
- Graduata
- Gamma di capacità disponibili
- Raccordi Luer Lock o Luer Slip
Specifica
Polypropylene | |
2 mL | |
Luer Lock |
Fisherbrand Focus on glassware and plasticware
Dimensione file: 66.4 MB
Offerte speciali
-
Diversi produttori
Chromatography and Chemicals Promotion
Explore our new promotion and get up to 70% off some of our most popular products.Codice sconto: 24127Scade: 2025-05-31
nuova offerta Save Up to 70%
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Fisherbrand™ Siringhe in PP in plastica, Luer Lock > Volume: 2mL
Spot an opportunity for improvement?Share a Content Correction