Learn More
Fisherbrand™ PTFE Adjustable Turbine Shaft Stirrer
Fisherbrand™ PTFE Adjustable Turbine Shaft Stirrers are for use with plain glass, metal or PTFE Shafts.
Marca: Fisherbrand™ 005.91275
Descrizione
- Total Chemical Resistance
- High Purity
- Temperature Range -200°C to +280°C
- Non contaminating
- Easy to clean
Specifica
Fisherbrand™ PTFE Adjustable Turbine Shaft Stirrer | |
12 mm | |
°° |
75 mm | |
1 |
Titolo del prodotto
Fisherbrand™ PTFE Adjustable Turbine Shaft Stirrer > Shaft: 12mm; Rotor: 75mm