Learn More
Fisherbrand™ PTFE Adjustable Turbine Shaft Stirrer
Fisherbrand™ PTFE Adjustable Turbine Shaft Stirrers are for use with plain glass, metal or PTFE Shafts.
Marca: Fisherbrand™ 005.90875
Descrizione
- Total Chemical Resistance
- High Purity
- Temperature Range -200°C to +280°C
- Non contaminating
- Easy to clean
Specifica
Fisherbrand™ PTFE Adjustable Turbine Shaft Stirrer | |
8 mm | |
°° |
75 mm | |
1 | |
PTFE |
Titolo del prodotto
Fisherbrand™ PTFE Adjustable Turbine Shaft Stirrer > Shaft: 8mm; Rotor: 75mm