Learn More
behr Labor-Technik™ behrotest™ R 104 S Soxhlet Extractor Unit
Complete compact extraction system with basic rack, heating device, holder, hoses and glassware (reaction flasks, extractor, Dimroth condenser for extraction).
Marca: behr Labor-Technik™ B00218425
Descrizione
- Includes sample package, extraction sleeves and boiling stones.
- behr equipment for Soxhlet extraction comply with the most various requirements of daily laboratory routines.
- With behr hydrolysis units, acid digestion is possible before extraction (determination of total fat content according to Weibull and Stoldt).
- Practical holders for condenser and extraction adapter for secure placement between extractions
- Condenser with screw connections
- Condenser with screw connections
- Practical holders for condenser and extraction adapter for secure placement between extractions
- With behr hydrolysis units, acid digestion is possible before extraction (determination of total fat content according to Weibull and Stoldt).
- behr equipment for Soxhlet extraction comply with the most various requirements of daily laboratory routines.
Specifica
Extractor Unit | |
250 mL | |
4 sample positions |
Titolo del prodotto
behr Labor-Technik™ behrotest™ R 104 S Soxhlet Extractor Unit > Capacity: 100mL; vessel: round flasks
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto