Learn More
SCAT™ HPLC Safety Caps, V2.0
I tappi di sicurezza V2.0 sono progettati per estrarre in sicurezza i solventi quando si lavora con qualsiasi sistema HPLC/UHPLC. Estraete i vostri solventi in modo sicuro da bottiglie e serbatoi senza essere esposti a gas pericolosi e vapori di solvente.
Marca: SCAT™ 307909-PP
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
SCAT™ Safety Caps V2.0 provide maximum safety for your HPLC solvent supply.
- High quality materials ensure maximum safety for lab and staff
- 360° rotatable and up to 200°C autoclavable screw cap for maximum flexibility
- Stable construction for highest safety and easy handling
- Chemically resistant against aggressive organic solvents
- Attractive design and clear arrangement of connections
- Improved design for easy connection of capillaries in various colors
- Free air valve with change indicator included with every Safety Cap
Specifica
HPLC Systems | |
Safety Cap II: 1 x air valve, 1 x change indicator, 2 x PP fitting 1.6/2.3/3.3 mm OD, 1 x PP blind plug, 1 x labelling field | |
GL45 |
Lab Bottle | |
Polypropylene | |
Safety Cap |
Titolo del prodotto
SCAT™ HPLC Safety Caps, V2.0 > Includes: Safety Waste Cap II: 1 x air valve, 1 x change indicator, 2 x PP fitting 1.6/2.3/3.3 mm OD, 1 x PP blind plug, 1 x labelling field, Material: PP, Thread Size: GL45