Estrazione e purificazione delle proteine
Thermo Scientific™ Soluzione detergente Triton X-100 Surfact-Amps™
Thermo Scientific™ Surfact-Amps X-100 Detergent Solution is highly-purified Triton X-100 detergent stabilized and supplied as a 10% solution in 10mL glass ampules.
Thermo Scientific™ Halt™ Protease and Phosphatase Inhibitor Cocktail (100X)
Prevent protein degradation and stabilize phosphorylation with this all-in-one cocktail of protease and phosphatase inhibitors.
Thermo Scientific™ Halt™ Protease Inhibitor Cocktail, EDTA-Free (100X)
Inhibit proteolytic degradation during cell lysis and protein extraction with these ready-to-use concentrated stock solutions of protease inhibitors.
Thermo Scientific™ Sodio desossicolato Pierce™
Water-soluble, ionic detergent used in cell lysis buffers (e.g., RIPA buffer) to disrupt protein interactions for liposome and membrane protein isolation.
Cytiva Colonne preimpaccate Superdex™ 200 Increase 10/300 GL Tricorn™
Per cromatografia ad esclusione dimensionale e analisi di proteine con pesi molecolare tra 10,000 e 600,000. Le colonne preimpaccate Cytiva Lifescience™ Superdex™ 200 Increase 10/300 GL Tricorn™ sono progettate per purificazione preparativa su piccola scala.
Thermo Scientific™ Kit di frazionamento subcellulare di proteine per cellule in coltura
Enrich for cultured cell-specific cytoplasmic, membrane, nuclear soluble, chromatin-bound and cytoskeletal proteins in less than 3 hours.
Thermo Scientific™ Kit di isolamento mitocondriale per cellule in coltura Pierce™
Isolate intact mitochondria from cultured mammalian cell samples in about 40 minutes with this versatile, benchtop microcentrifuge-based fractionation kit.
Thermo Scientific™ Reagente di estrazione di proteine da tessuto T-PER™
Extract total protein from tissue samples using this mild cell lysis and extraction reagent developed specifically for tissues.
Thermo Scientific™ Lisozima Pierce™
Increase protein or nucleic acid extraction efficiency with this enzyme that is characterized by the ability to break down the bacterial cell wall.
Thermo Scientific™ Resina HisPur™ Ni-NTA
Purify His-tagged recombinant fusion proteins using high-capacity nickel-NTA (nitrilotriacetic acid) metal-chelate beads, spin columns and plates.
Cytiva Colonne HiTrap™ Chelating HP
Purificazioni ottimali di proteine con tag di istidina quando gli ioni nichel non sono la scelta migliore. Le colonne Cytiva Life Sciences™ HiTrap™ Chelating HP sono preimpaccate con Chelating Sepharose™ ad alte prestazioni.
Thermo Scientific™ Pierce™ GST Protein Interaction Pull-Down Kit
Easily capture and affinity purify binding partners of GST-tagged fusion proteins with these glutathione (GSH) agarose beads and complete reagent set.
Thermo Scientific™ Pierce™ Streptavidin Agarose
Bind biotinylated proteins and other ligands with a wide selection of high-quality agarose and acrylamide beads and columns of recombinant streptavidin biotin-binding protein.
Thermo Scientific™ Soluzione detergente NP-40 Surfact-Amps™
Thermo Scientific™ Surfact-Amps NP-40 Detergent Solution is highly-purified NP-40 detergent stabilized and supplied as a 10% solution.
Thermo Scientific™ HisPur™ Ni-NTA Spin Columns, 3 mL
Purify His-tagged recombinant fusion proteins using high-capacity nickel-NTA (nitrilotriacetic acid) metal-chelate beads, spin columns and plates.
Thermo Scientific™ Tampone RIPA Pierce™
Lyse cultured mammalian cells with this high-quality, ready-to-use and fully disclosed formulation of a popular cell lysis reagent.
Sodium Dodecyl Sulfate (SDS), White Powder, Electrophoresis, Fisher BioReagents™
Formula molecolare: C12H25NaO4S Molecular Weight (g/mol): 288.38 Numero MDL: MFCD00036175 InChI Key: DBMJMQXJHONAFJ-UHFFFAOYSA-M Sinonimo: SDS, Sodium Lauryl Sulfate PubChem CID: 3423265 ChEBI: CHEBI:8984 IUPAC Name: sodium dodecyl sulfate SMILES: [Na+].CCCCCCCCCCCCOS([O-])(=O)=O
Thermo Scientific™ Resina per la rimozione di endotossine di elevata capacità Pierce™
Remove endotoxins (lipopolysaccharides or LPS) from samples before mass spectrometry using spin columns of poly(e-lysine) ligand on beaded cellulose.
Thermo Scientific™ Bigliemagnetiche alla streptavidina Pierce™
Purify or immunoprecipitate biotinylated proteins or affinity ligands using streptavidin-coated magnetic particles for magnet-based separation.
Thermo Scientific™ Inibitori della proteasi Pierce™
Customize your protease inhibitor cocktail formulation with these small packages of individual protease inhibitor peptides and compounds.
Thermo Scientific™ Pierce™ Protease Inhibitor Tablets, EDTA-free
Thermo Scientific™ Pierce Protease Inhibitor Tablets are broad-spectrum protease inhibitors that are highly effective at preventing proteolytic degradation for cell lysis and protein extraction experiments
Sartorius Vivaflow™ Models Accessories: Diafiltration Reservoir
For Vivaflow 50, 200
Sartorius™ Vivaflow™ 50R, 200 Accessory Set
Complete set of accessories for Vivaflow 50R, 200 with 230V pump.
Thermo Scientific™ Kit di estrazione nucleare e citoplasmatica NE-PER™
Efficiently lyse cells and extract separate cytoplasmic and nuclear protein fractions in less than two hours with this subcellular fractionation kit.
Thermo Scientific™ Cocktail inibitore della fosfatasi Halt™
Stabilize serine, threonine and tyrosine phosphorylation of proteins in cells and tissues with this cocktail of phosphatase inhibitors.
Thermo Scientific™ Triton™ X-114 Surfact-Amps™ Detergent Solution
Purified nonionic Triton X-114 detergent stabilized and supplied as a 10% solution in 10mL glass ampules.
Thermo Scientific™ Brij™-35 Surfact-Amps™ Detergent Solution
Stabilized Brij-35 detergent provided as 10% and 30% solutions for use in various protein methods including cell lysis and HPLC.
Thermo Scientific™ Pierce™ Glutathione Agarose
Purify GST-tagged recombinant fusion proteins from cell lysates using glutathione (GSH) beaded agarose affinity purification resins, spin columns and plates.
Thermo Scientific™ Lysozyme
Degrade unwanted bacterial cell wall in protein or nucleic acid extracts