Laboratory Freezers
Fryka™ TS Freezer
Fryka TS Freezer has usable space of 100L and can be used for cooling and freezing in industrial and scientific laboratories.
Fryka™ Cold Box
Reliable, compact, and quiet cold box designed as a desktop appliance. Fryka™ Cold Box facilitate decentralized storage of chilled items.
Thermo Scientific™ TSG General Purpose Freezers - Crated
The TSG Series of general purpose freezers are built with the performance, security, and quality features you have come to expect from Thermo Scientific™.
Thermo Scientific™ TSX Series High-Performance -20°C Manual Defrost Freezers
Thermo Scientific™ TSX Series high-performance -20°C manual defrost freezers offer a sustainable choice for clinical and laboratory storage
Liebherr™ Spark-Free Upright Lab Freezer with Shelves
Secure, accurate and ultra energy-efficient, spark-free interior, freezer storage, the Liebherr™ Spark-Free Interior, Lab Freezer is designed for general purpose applications with Liebherr build-quality you can rely on
Liebherr™ Upright Lab Freezer with Drawers
Secure, accurate and ultra energy-efficient freezer storage of temperature-sensitive products, the Liebherr™ Upright Lab Freezer is fully-equipped for lab applications with Liebherr build-quality you can rely on.
Liebherr™ Tall Heavy-Duty Lab Freezer
For secure, accurate and long-lasting freezer storage of temperature-sensitive products, the Liebherr™ Tall Heavy-Duty Lab freezer is fully-equipped for lab applications with Liebherr build-quality you can rely on. EN 60068 temperature mapping and SmartMonitoring options.
Fryka™ B 30 Cold Box
Compact and space saving. Fryka B 30 Cold Box designed as desktop appliances and are used for cooling and freezing for all research and industry laboratories.
Liebherr™ Upright Lab Freezer with Shelves
Secure, accurate and ultra energy-efficient freezer storage, the Liebherr™ Lab Freezer is designed for general purpose applications with Liebherr build-quality you can rely on.
Liebherr™ ProfiLine Forced-Air Freezer
Use these freezers for general laboratory storage of temperature-sensitive samples. Liebherr™ ProfiLine Forced-Air Freezers feature chrome-nickel-steel interior containers and plastics coated grid shelves.
Liebherr™ SmartFrost Comfort Freezer
Use these freezers for general laboratory storage of temperature-sensitive samples. Liebherr™ SmartFrost Comfort Freezers provide a more flexible storage area with greater capacity and improved energy consumption for lower running costs.
![](https://assets.fishersci.com/TFS-Assets/LED/product-images/F63860~p.eps-650.jpg)
Laboratory freezers are essential items of equipment in many laboratories, especially those that require the safe and secure short-term or long-term frozen storage of temperature-sensitive biological or chemical materials.
Lab freezers are offered as upright vertical, horizontal chest or under-bench units in a range of sizes and capacities. Small, compact benchtop models are ideal for personal use.
A general-purpose laboratory freezer typically stores samples down to -25°C. An ultra-low temperature (ULT) freezer is intended for deep cryogenic storage of samples typically down to -80°C.
Most freezers are built from robust and durable materials such as stainless steel, aluminium, or powder-coated sheet steel.
Laboratory freezers have a selection of internal storage layouts or compartments, some of which may be reconfigurable by the user. For example, they may have combinations of adjustable and exchangeable doors, shelves, baskets, trays or special racking systems designed for the storage of cryogenic boxes, microplates or cryotubes.
Laboratory freezers come equipped with a range of standard or additional features such as:
- Energy-efficient, microprocessor controlled refrigeration systems
- Digital, user-friendly touchscreen displays
- Lockable, card-entry or self-closing doors for extra safety and sample security
- Spark-free versions for storage of flammable
- Over-temperature audible or visual alarms
- Remote temperature monitoring with USB data download capability
- Inner doors and high-efficiency foam insulation to minimise temperature losses
- Highly efficient pump systems for fast cool-down times
- Fan-assisted forced-air circulating systems to remove moisture and minimise icing
- Automatic gas-powered defrost systems
- CO2 or LN2 safety back-up systems in the event of a power or mechanical failure