missing translation for 'onlineSavingsMsg'
Learn More
2-Chloro-3-cyanopyridine-4-boronic acid pinacol ester, 95%, Thermo Scientific™ Product Code.: 11332339

2-Chloro-3-cyanopyridine-4-boronic acid pinacol ester, 95%, Thermo Scientific™

Product Code. 11332339
1 g
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy

Product Code. 11332339

Brand: Thermo Scientific Alfa Aesar H27284.03

Request Bulk or Custom Format

Effettua il per acquistare questo articolo Hai bisogno di un conto web ? Registrati oggi stesso!

Questo articolo non è attualmente disponibile o è stato dismesso.
La preghiamo di consultare la pagina del prodotto per individuare le possibili alternative.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 878194-94-6
Molecular Formula C12H14BClN2O2
Molecular Weight (g/mol) 264.516
MDL Number MFCD09037479
InChI Key YYPZHORJYDCOSQ-UHFFFAOYSA-N
Synonym 2-chloro-3-cyanopyridine-4-boronic acid pinacol ester, 2-chloro-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl nicotinonitrile, 2-chloro-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl pyridine-3-carbonitrile, 2-chloro-3-cyano-4-pyridinylboronoic acid pinacol, 2-chloro-3-cyano-4-pyridinylboronic acid pinacol ester, 2-chloro-3-cyano-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl pyridine, 2-chloro-4-tetramethyl-1,3,2-dioxaborolan-2-yl pyridine-3-carbonitrile
PubChem CID 44755174
IUPAC Name 2-chloro-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine-3-carbonitrile
SMILES B1(OC(C(O1)(C)C)(C)C)C2=C(C(=NC=C2)Cl)C#N

Specifications

Melting Point 100°C to 101°C
Quantity 1 g
Formula Weight 264.52
Percent Purity 95%
Chemical Name or Material 2-Chloro-3-cyanopyridine-4-boronic acid pinacol ester
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
2-Chloro-3-cyanopyridine-4-boronic acid pinacol ester, 95%, Thermo Scientific™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.