Learn More
Abeba™ Classic 1500 Shoes
Animal fats, oil and fuel resistant. Leather/microfiber construction.
Marca: Abeba™ 1500/41
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
- CE EN ISO 20345:2011 S2 SRC
- Grained leather
- White
- Steel toe cap
- Slip resistance PU outsole sole
- Sanitized treated
- Antistatic
- Lining microfiber, respiration absorbing
- adjustable instep area
Specifica
CE EN ISO 20345:2011 S2 SRC | |
Microfiber | |
41 | |
Polyurethane | |
Steel |
White | |
Leather | |
Slip-resistant | |
1500 | |
Animal fats, oil and fuel |
Titolo del prodotto
Abeba™ Classic 1500 Shoes > Size: 41