Learn More
Memmert™ Entry Port 23 mm for UN30, Custom Position TBC Left Side
Optional entry port for Memmert Ovens.
Marca:  Memmert™ UN30F4
Codice prodotto. 17570086
  • 195.00€ / Pezzo
Spedizione stimata: 28-05-2025
per visualizzare lo stock disponibile
Esplora altre offerte speciali
Aggiungi al carrello
Questo articolo non è restituibile. Consulta la politica di reso
Descrizione
Descrizione
  • Entry port with 23 mm diameter
  • Flap closure
  • Custom position TBC in left side
Specifica
Specifica
UN30 Oven
Entry Port
23 mm
Suggerimenti di prodotto
Suggerimenti di prodotto
Videos
SDS
Documenti
Documenti
One moment while we fetch your results.
Certificati
Speciale aanbiedingen
Speciale aanbiedingen
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Memmert™ Entry Port 23 mm for UN30, Custom Position TBC Left Side > For Use With: UN30
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.