missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ Exosome component 5 Protein
Highly purified. Generating reliable and reproducible results.
Marca: Novus Biologicals™ NBP1-99063-100UG
This item is not returnable.
View return policy
Description
A recombinant protein with a N-Terminal His-tag and corresponding to the amino acids 1-235 of Human Exosome component 5 The Recombinant Human Exosome component 5 Protein is derived from E. coli. The Recombinant Human Exosome component 5 Protein has been validated for the following applications: SDS-Page.
Specifications
0.5mg/mL | |
Liquid. In PBS (pH 7.4) containing 2mM DTT, 30% glycerol, 200mM NaCl | |
SDS-PAGE | |
Human | |
RUO | |
Unconjugated |
SDS-PAGE | |
27.5kDa | |
100μg | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
Exosome component 5 | |
>90% |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Novus Biologicals™ Exosome component 5 Protein > 100μg
Spot an opportunity for improvement?Share a Content Correction