Learn More
Memmert™ Full-Sight Glass Door for UN260, Max. 250°C
Full-sight glass door constructed of four insulated glass layers.
Marca: Memmert™ UN260B0
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
- Accessory
- Full-sight
- Glass (4 layers)
- Temperature range up to 250°C
Specifica
UN260 Oven | |
250°C (Max.) | |
Full-sight Door | |
Glass |
Suggerimenti di prodotto
Customers who viewed this item also viewed.
Titolo del prodotto
Memmert™ Full-Sight Glass Door for UN260, Max. 250°C > For Use With: UN260