Learn More
Smeg™ Injection Trolley for Laboratory Glassware Washers
Use these trolleys with Smeg™ glassware washers. Smeg Injection Trolleys for Laboratory Glassware Washers are available in a variety of configurations to fit a wide range of applications.
Marca: Smeg™ LM40DS
Descrizione
- For use with Smeg glassware washers
- Available with one, two and mixed spraying/injection levels
- Clean and sanitize a wide variety of labware
Specifica
Stainless Steel | |
40 items | |
Injection trolley with one injection level | |
Laboratory glassware washers | |
1 Each |
Injection Trolley | |
Capacity of 40 items | |
Washing narrow-necked glassware up to 490mmH | |
40 spigots, drying system connection |
Suggerimenti di prodotto
I clienti che hanno visualizzato questo articolo hanno visto anche