Learn More
Fisherbrand™ PTFE Adjustable Turbine Shaft Stirrer
Fisherbrand™ PTFE Adjustable Turbine Shaft Stirrers are for use with plain glass, metal or PTFE Shafts.
Marca: Fisherbrand™ 005.910100
Descrizione
- Total Chemical Resistance
- High Purity
- Temperature Range -200°C to +280°C
- Non contaminating
- Easy to clean
Specifica
Fisherbrand™ PTFE Adjustable Turbine Shaft Stirrer | |
10 mm | |
°° |
100 mm | |
1 |