Learn More
Eppendorf™ RackScan
RackScan reads complete storage racks with up to 96 different barcoded vials. The sample IDs are transferred to downstream databases.
1595.00€ - 10845.00€
Codice del prodotto | Marca | Tipo | Prezzo | Quantità e disponibilità | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Codice del prodotto | Marca | Tipo | Prezzo | Quantità e disponibilità | ||||||
16476879
|
Eppendorf™
6002000006 |
Scanner (Bottom Reading) |
10845.00€
Pezzo |
In stock
Eseguire il login per visualizzare lo stock disponibile |
||||||
16486879
|
Eppendorf™
6002000014 |
Scanner (Side Reading) |
1595.00€
Pezzo |
In stock
Eseguire il login per visualizzare lo stock disponibile |
||||||
Descrizione
- Camera based instrument resulting in rapid imaging and decoding
- Comfortable and easy plug-in accessory
- Small footprint, ideal for benchtop (and integration)
- ECC 200 error correction function enables safe code reading with up to 30 % damaged codes
- Support material of vials available via Eppendorf Homepage for documentation purposes
- Plug & Transfer with Eppendorf™ eLABInventory and eLABJournal software for comfortable data transfer
- RackScan s can be attached to RackScan b as accessory to read barcodes on side labels
- Different formats usable for high flexibility
Specifica
PC/USB | |
USB |
Titolo del prodotto
Eppendorf™ RackScan