Learn More
Watson-Marlow Manifold Tubing - PVC
Double pump segment manifold tubing — use one, transfer to the other for double life
Marca:  Watson-Marlow 981.0038.000
Codice prodotto. 10782866
  • 169.00€ / Pezzo
Spedizione stimata: 07-04-2025
per visualizzare lo stock disponibile
Esplora altre offerte speciali
Aggiungi al carrello
Descrizione
Descrizione
Double pump segment manifold tubing — use one, transfer to the other for double life
  • For use with 400DM2 and 400DM3 pumpheads
  • Can be used with any multichannel cartridges
  • Adjacent cartridges may contain tubing of a different material or size
TRUSTED_SUSTAINABILITY
Specifica
Specifica
7/500 in.
PVC
0.38 mm
0.8 mm
0.38mm
Orange/Green
0.38 mm
0.014 in.
0.03 in.
323S/MC peristaltic pump
0.013 to 0.92 (four roller pump), 0.012 to 0.830 (eight roller pump)mL/min.
Pump Tubing
Titolo del prodotto
Watson-Marlow Manifold Tubing - PVC > Material: PVC; Bore: 0.38mm; Color code: Orange/green