Siringhe per cromatografia
Thermo Scientific™ Siringhe GC per autocampionatore TriPlus™ RSH
Iniezioni accurate, precise e riproducibili con le siringhe GC Thermo Scientific™ per autocampionatore TriPlus™ RSH Thermo Scientific™.
Thermo Scientific™ Fixed Needle Syringes for GC Instruments
Get accurate, reproducible results from these fixed-needle syringes for Thermo Scientific™ and Agilent GC autosamplers.
Hamilton™ Siringhe Gastight™ serie 1700: Terminale N
Versione Gastight delle siringhe originali serie 700, ideali per l'erogazione di volumi da 1 a 500 μl
Hamilton™ Siringhe Gastight™ serie 1000: Siringhe Luer-Lock, terminale TLL
Mid-volume GASTIGHT syringe ideal for dispensing volumes from 100 μL up to 100 mL.
Hamilton™ Siringhe Microliter™ serie 700: Terminale N
Siringa originale Hamilton assemblata a mano.
Hamilton™ Siringhe serie 7000 Modified Microliter™
This series of syringes is ideal for dispensing volumes from 0.05 µL up to 5 µL. The 7000 series is the only zero dead volume syringe. The fine gauge plunger wire travels to the tip of the needle ensuring that all solvent is expelled from the needle.
Hamilton™ Aghi a cono Kel-F a punta tagliata (Luer-Lock)
Kel-F hub (KF) needles can be used with Luer Tip (LT) and PTFE Luer Lock (TLL) syringes and connectors. Standard needle length is 2 in. (51mm). Custom lengths and point styles are available.
Hamilton™ Siringhe Gastight™ serie 1000: Siringhe ad ago fisso, terminale LTN, misura 22, lunghezza ago 2 poll.
Mid-volume GASTIGHT syringe ideal for dispensing volumes from 100 μL up to 100 mL. A variety of different terminations and needle options are available in this series.
Hamilton™ 1000 Series Gastight™ Syringes: Luer Lock Syringes, TLL Termination (No Flange)
Mid-volume GASTIGHT syringe ideal for dispensing volumes from 100 μL up to 100 mL.
Hamilton™ Siringhe Gastight™ serie 1700: Terminale RN
Gastight version of the original 700 series syringes, ideal for dispensing volumes from 1 up to 500μL
Hamilton™ Siringhe serie 700/1700 Microliter/Gastight™
Hamilton Company offre la selezione più completa del mercato di siringhe standard e personalizzate. Ciascuna di esse è realizzata per offrire il massimo livello di precisione e accuratezza possibile.
Hamilton™ Siringhe Gastight™ serie 1700: Terminale TLL
Gastight version of the original 700 series syringes, ideal for dispensing volumes from 1 up to 500μL
Hamilton™ 1700 and 1000 Series Gastight™ SampleLock™ Syringes, 22 Gauge
Hamilton SampleLock syringes allow you to collect, store, transport and analyze liquid or gaseous samples without the danger of evaporative loss or environmental contamination.
Trajan™ Carlo Erba On-Column Syringe
On-column syringes require the needle OD to be smaller than the ID of the capillary column with which it is used. For use with Carlo Erba products
Thermo Scientific™ Manual GC Syringes
Easily accomplish accurate, precise, and reproducible manual injections with Thermo Scientific™ GC Syringes.
Hamilton™ Siringhe Gastight™ serie 1700: Terminale LTN
Gastight version of the original 700 series syringes, ideal for dispensing volumes from 1 up to 500μL
Hamilton™ 700 and 1700 Series Gastight™ and Microliter™ Digital Syringes: N Termination
Easily achieves precision up to 0.5% while dispensing volumes from 500μL down to 50nL
SGE™ Siringhe NanoVolume
Ideal for nanovolume capillary chromatography injection as well as preparing accurate standards which require small volumes.
Hamilton™ Siringhe per autocampionatori PAL LC Microliter™ e Gastight™ con tecnologie CTC e LEAP
Per l'uso con CTC LC PAL, HTC PAL, HTS PAL HTX PAL, PAL HTC-xt, PAL HTS-xt, PAL HTX-xt estesi e altri sistemi di autocampionamento
Thermo Scientific™ Removable-Needle Autosampler Syringes for GC Instruments
Thermo Scientific™ Removable-Needle Autosampler Syringes for GC Instruments are accurate and reproducible over a range of volumes.
Hamilton™ Siringhe Microliter™ serie 700: Terminale RN
Siringa originale Hamilton assemblata a mano.
Trajan™ Jumbo Syringe
Hold and dispense large gas volumes. Lightweight and ideal for medical equipment calibration.
Trajan™ Syringe Plunger and Needle Kits
Designed to inject down to 1mL with high precision and accuracy
Hamilton™ e siringhe con ago cementato GC per autocampionatori Gastight™ Agilent 7670, 7671 e 7672 ALS
Per gli autocampionatori di liquidi per gascromatografia Agilent 7670, 7671 e 7672 Automatic Liquid Sampler (ALS). Le siringhe con ago cementato GC per autocampionatori ALS Gastight™ Agilent 7670, 7671 e 7672 hanno l'estremità dello stantuffo in PTFE che aderisce perfettamente al cilindro della siringa, consentendo il campionamento di liquidi o gas.
SGE™ Siringhe per autocampionatore
La tecnologia Diamond delle siringhe offre maggiore resistenza ai solventi, ampie gamme di temperature e una migliore fluidità operativa. Le siringhe per autocampionatori SGE PAL con ago fisso a punta conica da 50 mm sono utilizzate con autocampionatori CTC, LEAP, CombiPAL, GC PAL, Thermo TriPlus, AS2000, AS3000, AS200/800.
Hamilton™ 700 7000/1700 1000 Series Microliter™/Gastight™: TLL Termination
Syringes have built-in Chaney Adapters
Hamilton™ Aghi a cono amovibili grandi (siringhe da 250 μl - 10 ml)
Gli aghi a cono rimovibili grandi si possono utilizzare con siringhe ad ago rimovibile tipo (RN) da 250 μl e superiori. La lunghezza standard dell'ago è 2 inch (51 mm). Sono disponibili misure, lunghezze e tipi di punta personalizzati.
SGE™ Siringhe a stantuffo guidato
Extended barrel guides plunger during injection. SGE™ Guided Plunger Syringes are suitable for industrial environments.
Hamilton™ Siringhe Gastight™ serie 1000: Ago amovibile, terminale RN, misura 22, lunghezza ago 2 poll.
La siringa GASTIGHT di volume medio è ideale per l'erogazione di volumi compresi tra 100 μl e 100 ml.