Controlled Environments Bags
LABPLAS TWIRL’EM Clear Safety Tabs
Clear Safety Tabs can accommodate solid, semi-solid and liquid samples.
Micronova™ Brush Covers
These 100% smooth polyester brush covers work with the Cylinder and Kettle Brushes to improve fine cleaning and sanitization of small tanks, carboys and other process equipment. Made with FDA approved material for indirect food contact.
Keystone Cleanroom Products Gamma-Irradiated Heat-Seal Sterility Maintenance Bag
Bag used to cover wrapped items after sterilization to provide protection from environmental factors that could compromise sterility.
Keystone Cleanroom Products™ Super Duty SMS Autoclavable Bowl Cover
This 88 g/m2 SMS Autoclavable Bowl Cover provides a microbial barrier ideal for pharmaceutical production applications.
Keystone Cleanroom Products™ Blue Heavy Duty Autoclavable SMS Muff Cover
This 50 g/m2 SMS Autoclavable Muff Cover provides a microbial barrier ideal for pharmaceutical production applications.
Micronova™ Irradiated BucketLiners™
Creates an impermeable barrier to strong solutions and raw materials when used to line cleanroom buckets and storage containers.
Keystone Cleanroom Products™ Blue Super Duty Autoclavable SMS Muff Cover
This 88 g/m2 SMS Autoclavable Muff Cover provides a microbial barrier ideal for pharmaceutical production applications.
Keystone Cleanroom Products Gamma-Irradiated 2 mil Polyethylene Trash Bag
Simplify the removal of waste from your aseptic cleanroom without introducing external sources of microbial contamination, by using gamma irradiated trash liners.
Keystone Cleanroom Products™ Heavy Duty SMS Autoclavable Bowl Cover
This 50 g/m2 SMS Autoclavable Bowl Cover provides a microbial barrier ideal for pharmaceutical production applications.
Contec™ Disposal Bag for Mop Removal Tool
Disposal bag for mop head removal tool
Contec™ Rivestimenti sterili per secchi in polietilene
Compatibile con secchi da 5 galloni (20 l), 6.5 galloni (25 l) e 7 galloni (27 l). I rivestimenti sterili per secchi Contec™ proteggono i secchi e le soluzioni disinfettanti riducendo la possibilità di contaminazione incrociata.
Micronova™ Hazard Bags
These low density, LDPE hazard bags are designed for containing biomedical and hazardous waste in aseptic and controlled environments.
Micronova™ Sacchi per lo smaltimento dei rifiuti BinLiners
These FDA compliant, Sterile LDPE bags are designed to contain and transport biomedical waste in aseptic environments. Available with thicknesses ranging from 1 - 6 mil, and capacity to 60 gal. (200 L)
Gilac™ Container Cover
Container Cover is coated with the Cleanbake™ technology, a proven-effective textile solution that reduces moisture and prevents the build up of mildew, dirt and dough residues.
Keystone Cleanroom Products BHD Blue Autoclaveable Wrapper
When secured with autoclave tape or elasticized bands, Keystone Cleanroom Product’s Blue BHD wrappers provide an adequate microbial barrier from external sources of contamination.
Contec™ Irradiated Disposal Bag for Mop Removal Tool
Irradiated disposal bag for mop head removal tool
Micronova™ Brush Covers
These 100% smooth polyester brush covers work with the Cylinder and Kettle Brushes to improve fine cleaning and sanitization of small tanks, carboys and other process equipment. Made with FDA approved material for indirect food contact.
Kolmi Hopen™ White Polyethylene Valve Cover
This versatile valve cover can be used to cover containers, jerrycans, valves, bins, buckets, and more.
Micronova™ BinLiners™
These FDA compliant, LDPE bags are designed for cleanroom material disposal and transfer, garment collection and equipment covers in controlled environments. Available with thicknesses ranging from 1 - 6 mil, and capacity to 60 gal. (200 L).