Learn More
BD Cytometric Bead Array (CBA) Human IL-10; Flex Set, Bead B7
Bead-based immunoassay capable of measuring human interleukin-10 (IL-10) in serum, plasma, and cell culture supernatant samples
Brand:  BD 558274
Product Code. 15809278
  • 670.00€ / Pezzo
Explore more special offers
Includes: Human IL-10; Standard; Human IL-10; PE Detection Reagent; Human IL-10; Capture Bead B7
This item is not returnable. View return policy
Description
Description
Human reactivity was determined by testing samples with the BD CBA Human IL-10 Flex Set
Flow cytometry
TRUSTED_SUSTAINABILITY
Specifications
Specifications
IL-10
Human IL-10, standard, Human IL-10, PE detection reagent, Human IL-10, capture bead B7
Serum, Plasma, Cell Culture Supernatant
Flow Cytometry
Human
100 Tests
4°C
Human IL-10 flex set (bead B7)
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
BD Cytometric Bead Array (CBA) Human IL-10; Flex Set, Bead B7 > Human IL-10; 100 Tests
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.
Research Use Only