missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD3/CD28 Antibody Cocktail, FITC, PE, Invitrogen™
Antibody Cocktail
Marca: Invitrogen MA112098
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
This antibody is a monoclonal mixture of FITC-conjugated CD3 (clone M2AB, isotype IgG1) and PE-conjugated CD28 (clone B-23, isotype IgG1).
T cell activation through the antigen receptor (TCR) involves the cytoplasmic tails of the CD3 subunits CD3 gamma, CD3 delta, CD3 epsilon and CD3 zeta. These CD3 subunits are structurally related members of the immunoglobulins super family encoded by closely linked genes on human chromosome 11. The CD3 components have long cytoplasmic tails that associate with cytoplasmic signal transduction molecules. This association is mediated at least in part by a double tyrosine-based motif present in a single copy in the CD3 subunits. CD3 may play a role in TCR-induced growth arrest, cell survival and proliferation. Human CD28 antigen is a 44 kDa disulfide linked homodimeric T cell specific surface glycoprotein. It is a cell adhesion molecule of the immunoglobulin superfamily which is constitutively expressed on most peripheral blood T lymphocytes (approximately 95% of CD4+ cells and 50% of CD8+ cells). Mature thymocytes exhibit higher levels of CD28 than the immature cells. Activation of T cells results in enhanced CD28 expression. T cell activation requires two combined signals provided by antigen presenting cells.
Specifica
CD3/CD28 | |
Cocktail | |
PE | |
CD28 | |
CD28; CD28 antigen; CD28 molecule; CD3 antigen, delta subunit; CD3 delta; CD3 epsilon; CD3D; CD3d antigen, delta polypeptide (TiT3 complex); CD3d molecule; CD3d molecule, delta (CD3-TCR complex); CD3-DELTA; CD3E; CD3e antigen, epsilon polypeptide (TiT3 complex); CD3e molecule; CD3e molecule, epsilon (CD3-TCR complex); CD3-epsilon; CD3G; CD3g antigen, gamma polypeptide (TiT3 complex); CD3g molecule; CD3g molecule, epsilon (CD3-TCR complex); CD3g molecule, gamma (CD3-TCR complex); CD3-GAMMA; FLJ18683; IMD17; IMD18; IMD19; MGC138290; OKT3, delta chain; T3D; T3E; T3G; T-cell antigen receptor complex, epsilon subunit of T3; T-cell antigen receptor complex, gamma subunit of T3; T-cell receptor T3 delta chain; T-cell receptor T3 gamma chain; T-cell surface antigen T3/Leu-4 epsilon chain; T-cell surface glycoprotein CD3 delta chain; T-cell surface glycoprotein CD3 epsilon chain; T-cell surface glycoprotein CD3 gamma chain; T-cell-specific surface glycoprotein CD28; TCRE; Tp44 | |
Mouse | |
Protein A/G | |
RUO | |
915, 916, 917, 940 | |
4° C, store in dark, DO NOT FREEZE! | |
Liquid |
Flow Cytometry | |
M2AB, B-23, B-23 | |
PBS with 0.2% carrier protein and 0.08% sodium azide | |
P04234, P07766, P09693, P10747 | |
CD28, CD3D, CD3E, CD3G | |
human thymocytes and HPB-ALL T-cell line | |
50 Tests | |
Primary | |
Human | |
Antibody | |
IgG1 |
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto
CD3/CD28 Antibody Cocktail, FITC, PE, Invitrogen™ > 50 Tests; FITC, PE
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto