Learn More
Value Plastics Female Luer to 200 Series Barb
Luer tapers that are constructed to meet ISO 594-1 and 594-2 standards
Marca: Value Plastics 50120-45
Descrizione
Rotating locks minimize twisting and kinking of tubing during positioning. Tapered cone seals are reliable and uncomplicated. Facilitate single-use configurations that minimize downtimeSpecifica
ADCF;FDA;Non-DEHP;Non-Phthalate;RoHS;USP | |
Polypropylene | |
Female Luer x Hose Barb |
5/32 in. | |
25/Pk. |
Always test the application to verify the performance of the interconnection.