missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Goat anti-Chicken IgG (H+L) Secondary Antibody, Biotin, Novus Biologicals™
Goat Polyclonal Antibody
Marca: Bio-Techne NB7587
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
IgG (H+L) Polyclonal specifically detects IgG (H+L) in Chicken samples. It is validated for Western Blot, ELISA, Immunohistochemistry.
Specifica
IgG (H+L) | |
Polyclonal | |
Western Blot 1:3000 - 1:30000, ELISA 1:800000, Immunohistochemistry 1:500 - 1:3000, Dot Blot | |
Goat | |
Immunogen affinity purified | |
RUO | |
Secondary | |
Chicken | |
Purified |
Western Blot, ELISA, Immunohistochemistry, Dot Blot, Immunofluorescence, Immunohistochemistry (Paraffin) | |
Biotin | |
IGHG | |
Chicken IgG whole molecule | |
1 mg | |
Adaptive Immunity, Immunology | |
This antibody will react with heavy chains of Chicken IgY and with light chains of most Chicken immunoglobulins. | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
IgG |
Suggerimenti di prodotto
Customers who viewed this item also viewed.
Viewing 1-4 of
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto
Goat anti-Chicken IgG (H+L) Secondary Antibody, Biotin, Novus Biologicals™ > 1mg; Biotin
For Research Use Only
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto