Learn More
Smeg™ Injection Trolley for Laboratory Glassware Washers
Use these trolleys with Smeg™ glassware washers. Smeg Injection Trolleys for Laboratory Glassware Washers are available in a variety of configurations to fit a wide range of applications.
Marca: Smeg™ LM40SDS
Descrizione
- For use with Smeg glassware washers
- Available with one, two and mixed spraying/injection levels
- Clean and sanitize a wide variety of labware
Specifica
Stainless Steel | |
Washing narrow-necked glassware up to 225mmH | |
40 U4140 spigots, drying system connection |
40 items | |
Laboratory glassware washers | |
1 Each |
Suggerimenti di prodotto
I clienti che hanno visualizzato questo articolo hanno visto anche
Accessori e Servizi
-
Smeg™ Carrello rack basso CARRELLO INFERIORE PER ACCESSORIMaglie l... Codice del prodotto: 11925614
328.00€ / Pezzo