Learn More
Kartell™ K70 Silicon Hoses
Kartell™ K70 Silicon Hoses are constructed of silicon, making them a transparent and flexible connection to various components and accessories.
Marca: Kartell™ 0392000
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
Kartell™ K70 Silicon Hoses are constructed of silicon, making them a transparent and flexible connection to various components and accessories.
- Flexible and transparent
- FDA, USP, Class VI, A 50p60, EPC VI.1.3.2 approved
- Can withstand temperatures of -60°C to 200°C
Specifica
FDA, USP, Class VI, A 50p60 and EPC VI.1.3.2 Approved | |
Silicone | |
10 m | |
10 m | |
200°C |
1 mm | |
3 mm | |
-60°C | |
Trasparente |
Titolo del prodotto
Kartell™ K70 Silicon Hoses > Inner Diameter: 1 mm