Learn More
Hamilton™ Long Life Instrument Syringes
Long life syringes were developed for extreme conditions that include harsh chemicals as well as abrasive particulate matter.
Marca:  Hamilton™ 8300-45
Codice prodotto. 10422385
  • 343.00€ / Pezzo
Esplora altre offerte speciali
Questo articolo non è restituibile. Consulta la politica di reso
Descrizione
Descrizione
  • Plunger tip made from UHMWPE (Ultra High Molecular Weight Poly Ethylene) which is harder than PTFE to better resist scratching
  • Plunger rod made from 316 stainless steel which offers exceptional chemical resistance
Specifica
Specifica
Borosilicate Glass, PTFE, UHMWPE
1/Ea.
Luer Lock
Instrument Syringe
5 mL
Documenti
One moment while we fetch your results.
Titolo del prodotto
Hamilton™ Long Life Instrument Syringes > Capacità: 5mL