Learn More
Thermo Scientific™ Nalgene™ Wide-Mouth Opaque Amber HDPE Bottles with Closure: Bulk Pack

Descrizione
Store, ship and pack light-sensitive reagents in Thermo ScientificTM NalgeneTM Wide-Mouth Opaque Amber HDPE Bottles with Closure.
- Opaque amber HDPE Bottles
- Opaque amber polypropylene closures
- Uniform walls resist splitting and puncturing
- Wide mouth allows easy filling and dispensing
- Bulk packed
- Leakproof†
Ordering Information:
Bottles and closures are packed in separate polybags within the same carton. † The term “leakproof” applies to Thermo Scientific Nalgene products that meet the following criteria: a) Bottle/flask/funnel closure systems with closures smaller than 100mm, after they are filled with water, inverted, withstand air pressure of 2psig for 2 minutes, and no water escapes;. b) Bottle/funnel/flask closure systems of larger than 100mm after they are filled with water, inverted for 15 minutes, and no water escapes. Note: these tests, using other liquids, may not yield the same results. To ensure safe usage, customers are advised to test Thermo Scientific Nalgene bottles and closures under conditions of their planned applications. Thermo Scientific Nalgene products are leakproof at ambient temperature and pressure when used with their Nalgene closures.
- I vantaggi per l'ambiente includono:
- Energia rinnovabile

Specifica
Specifica
Capacità | 8 once, 250 ml |
Capacità (sistema metrico) | 250 mL |
Capacità (sistema imperiale) | 8 once |
Colore | Opaque Amber |
Descrizione | Wide-Mouth Bottle |
Diametro (sistema metrico) interno collo | 33 mm |
Materiale | HDPE |
N. per custodia | 250 |
N. per confezione | 250 |
Sterilità | Non sterile |
Vedi altri risultati |
Suggerimenti di prodotto
I clienti che hanno visualizzato questo articolo hanno visto anche
Certificati
Certificati
Per poter ottenere i certificati è necessario indicare un numero di lotto. Per individuare il numero di lotto su ordini precedenti usi l'area dedicata allo stato degli ordini.
Numero di lotto | Tipo di certificato | Data | Catalog Number |
---|---|---|---|
1301731 | Certificato di conformità | 10/12/2024 | 312106-0008 |
1395372 | Certificato di conformità | 03/11/2023 | 312106-0008 |
1387578 | Certificato di conformità | 25/06/2023 | 312106-0008 |
1366332 | Certificato di conformità | 22/08/2022 | 312106-0008 |
1355852 | Certificato di conformità | 19/08/2022 | 312106-0008 |
† The term leakproof applies to Thermo Scientific Nalgene products that meet the following criteria: a) Bottle/flask/funnel closure systems with closures smaller than 100mm, after they are filled with water, inverted, withstand air pressure of 2psig for two minutes, and no water escapes;. b) Bottle/funnel/flask closure systems of larger than 100mm after they are filled with water, inverted for 15 minutes, and no water escapes. Note: these tests, using other liquids, may not yield the same results. To ensure safe usage, customers are advised to test Thermo Scientific Nalgene bottles and closures under conditions of their planned applications. Thermo Scientific Nalgene products are leakproof at ambient temperature and pressure when used with their Nalgene closures.
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.