Learn More
3M™ Particulate Respirator, FFP3, Valved, 8833SP
Cup-shaped disposable respirator with a comfortable M-shaped nose clip, a soft waffle edge, Cool Flow™ valve.
Marca:  3M™ 7100091323
18.80 EUR valido fino al 2025-06-08
Usa il codice promo "24131" per avere il tuo prezzo promozionale.
Codice prodotto. 15759817
  • 39.00€ / pezzi 5
Spedizione stimata: 07-05-2025
per visualizzare lo stock disponibile
Edge
Esplora altre offerte speciali
Aggiungi al carrello
Descrizione
Descrizione
  • Provides FFP3 (NPF 50) protection against dusts, oil-based mists and other fine particles
  • Robust, cup-shaped design
  • 3M™ Cool Flow™ Valve helps reduce heat build-up, even in hot and humid conditions
  • Features a full-face seal for improved fit and soft, cushioned lining for long-lasting comfort
  • Unique, M-shaped nose clip for extra customisation of fit
  • Lightweight, twin-strap design, nose foam, and soft waffle edge helps fit comfortably to a wide range of face sizes
  • Advanced Electret (filter) Media helps wearers breathe easily
  • Available in large or small pack sizes
  • The soft inner face-seal ring improves face seal and feeling of comfort and can be hygienically cleaned if used for longer than one shift (use 3M™ 105 Face Seal Cleaner)
  • Braided headbands are coloured for easy identification: red for FFP3
TRUSTED_SUSTAINABILITY
Specifica
Specifica
EN 149:2001 + A1:2009 FFP3 R D, CE Approved
5/Pk., 4 Pk./Cs.
No
General Purpose Particulate Respirator
Non-sterile
Offerte speciali
  • Diversi produttori
    Save up to 60% on gloves, apparel, and more
    Codice sconto: 24131
    Scade2025-06-08
    nuova offerta
    Shop Now
Titolo del prodotto
3M™ Particulate Respirator, FFP3, Valved, 8833SP > Quantity: 5/Pk., 4 Pk./Cs.