Learn More
DiscoverX PathHunter™ CHO-K1 cyno GPR120 β-Arrestin Cell Line
The PathHunter™ CHO-K1 cyno GPR120 β-Arrestin Cell Line measures GPR120 (GPCR) activity via recruitment of β-Arrestin2. This cell line ships as two vials of cryopreserved cells. Cyno protein.
Marca: DiscoverX 93-1029C2
Descrizione
The PathHunter™ cyno GPR120 β-Arrestin cell line is a stable, engineered cell line for use in studying drug candidates (small molecules or biologics) targeting GPCRs. This cell line enables assessment of ligand (e.g. free fatty acids) based activation of GPR120 GPCR activity via detection of β-arrestin recruitment. This product uses EFC technology (www.discoverx.com/technologies-platforms/enzyme-fragment-complementation-technology). The included cell line overexpresses PK-tagged cyno GPR120 and EA-tagged β-Arrestin 2. Activation of GPR120 stimulates the recruitment of β-Arrestin 2 and produces EFC signal. This stable modified cell line is provided as two vials of cryopreserved cells. Please note that this product expresses the Cyno version of the protein.Specifica
β-Arrestin Recruitment | |
Cell Lines | |
2 Vials | |
NM_001287319 |
CHO-K1 | |
GW9508 | |
cyno GPR120 |
Il programma Encompass di Fisher Scientific offre articoli che non fanno parte del nostro portafoglio di distribuzione. Questi prodotti in genere non hanno immagini o descrizioni dettagliate. Tuttavia, ci impegniamo a migliorare la vostra esperienza di acquisto. Si prega di utilizzare il modulo sottostante per fornire un feedback relativo al contenuto di questo prodotto.