missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RPL3 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marca: Invitrogen PA551912
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
Immunogen sequence: EDGKKQLEKD FSSMKKYCQV IRVIAHTQMR LLPLRQKKAH LMEIQVNGGT VAEKLDWARE RLEQQVPVNQ VFGQDEMIDV IGVTKGKGYK GVTSRWHTKK LPRKTHR Highest antigen sequence identity to the following orthologs: Mouse - 96%, Rat - 96%.
This gene encodes a member of the SWI2/SNF2 family of proteins, which play a critical role in altering protein-DNA interactions. The encoded protein has been shown to have dsDNA-dependent ATPase activity and RNA polymerase II termination activity. This protein interacts with cell division cycle 5-like, associates with human splicing complexes, and plays a role in pre-mRNA splicing.
Specifica
RPL3 | |
Polyclonal | |
Unconjugated | |
RPL3 | |
60S ribosomal protein L3; ASC-1; F2; HIV-1 TAR RNA-binding protein B; J1; J1 protein; L3; L4; Large ribosomal subunit protein uL3; OK/SW-cl.32; ribosomal protein L3; ribosomal protein L3 homolog; ribosomal protein L3, isoform a; RL3; RPL3; TARBP-B | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
6122 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunohistochemistry, Western Blot | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
P39023 | |
RPL3 | |
Recombinant protein corresponding to Human RPL3 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Suggerimenti di prodotto
Customers who viewed this item also viewed.
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto
RPL3 Polyclonal Antibody, Invitrogen™ > 100 μL; Unconjugated
Research Use Only
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto