missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ST2 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marca: Invitrogen PA523316
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
ST2 is a member of the interleukin 1 receptor family. Studies of ST2 in mouse suggested that this receptor can be induced by proinflammatory stimuli, and may be involved in the function of helper T cells. ST2 belongs to the interleukin-1 receptor family with three Ig-like C2-type (immunoglobulin-like) domains and a TIR domain. ST2 is a receptor for interleukin-33 (IL-33) which upon stimulation recruits the signaling molecules MYD88, IRAK1, IRAK4, and TRAF6, followed by phosphorylation of MAPK3/ERK1 and/or MAPK1/ERK2, MAPK14, and MAPK8. Reports also suggest its possible role in helper T-cell function as well as in cell growth. Three isoforms of ST2 have been reported where isoform C is a membrane form and isoform B is a secreted form. ST2 may be induced by proinflammatory stimuli, and may also be found in serum of autoimmune disease. Different isoforms are restricted to different tissues while high expression of this protein is evident in kidney, lung, placenta, stomach, skeletal muscle, colon and small intestine.
Specifica
ST2 | |
Polyclonal | |
Unconjugated | |
Q01638, P14719, Q62611 | |
IL1RL1 | |
Synthetic peptide (36 CPRQGKPSYTVDWYYSQT 53) of ST2/IL1RL1 protein. | |
100 μg | |
Primary | |
Human, Mouse, Non-human Primate, Rat | |
Antibody | |
IgG |
Immunohistochemistry (Paraffin), Western Blot | |
1.0 mg/mL | |
IL1RL1 | |
DER4, FIT-1, IL33R, ST2, ST2L, ST2V, T1, growth stimulation-expressed, homolog of mouse growth stimulation-expressed, interleukin 1 receptor-related protein, interleukin-1 receptor-like 1 | |
Rabbit | |
Protein G | |
RUO | |
17082, 25556, 9173 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Suggerimenti di prodotto
Customers who viewed this item also viewed.
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto
ST2 Polyclonal Antibody, Invitrogen™ > 100 μg; Unconjugated
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto