Controlled Environments Swabs
Texwipe™ Tampone Low TOC Alpha™
Constructed to exacting and consistent tolerances without the use of adhesives for superior performance. Texwipe™ Low TOC Alpha™ Swabs are cleeaned using a proprietary process to ensure suitability for use in Total Organic Carbon (TOC) analysis as part of a cleaning validation protocol.
Texwipe™ Tamponi in poliestere CleanTip™ serie Alpha™
Knitted and cleanroom laundered polyester head provides a neutral background for obtained sample because of its high cleanliness levels. Texwipe™ Alpha™ Swabs are widely recommended for surface sampling and cleaning validation work.
Texwipe™ CleanTip™ Swabs: CleanFoam™ Series
Excellent sorbency
Fisherbrand™ PurSwab Foam Swabs
Flexible foam applicators. Tip is comprised of an open-cell polyurethane foam. Ideal for a variety of cleaning applications.
Contec™ CONSTIX™ Surface Validation Swabs
Contec™ CONSTIX™ Surface Validation Swabs designed for the evaluation of surface cleaning protocols in biopharmaceutical industries.
Contec™ Tamponi in schiuma sigillati CONSTIX™
Sono disponibili diversi tamponi resistenti ideali per la pulizia. I tamponi sigillati in schiuma Contec™ CONSTIX™ sono eccellenti per la rimozione delle particelle. Per l’uso con solventi quali alcol isopropilico.
Texwipe™ Sterile Cotton Swabs
Made of 100% USP-grade cotton
Berkshire™ Nonwoven Polyester Lab-Tips™ Swabs
Nonwoven Polyester Swabs are available with small and long handles.
Texwipe™ CleanTip™ Swabs: Specialty Series
Certified to be silicone free
Contec™ Tamponi in poliestere sigillati CONSTIX™
Realizzati in tessuto di poliestere per camera sterile Polynit o tessuto di poliestere non tessuto per livelli eccezionalmente bassi di residui e particelle. I tamponi sigillati in poliestere Contec™ CONSTIX™ sono ideali per l'uso con solventi aggressivi quali acetone, triclorometano e acidi nitrico e solforico.
Texwipe™ Sterile Dry Collection and Transport System
Sterile ready-to-use units for the collection and transport of bacteriological specimens.
Contec™ Tamponi in cotone CONSTIX™
Realizzati con fibre di cotone cucite più lunghe e un semplice legante metilcellulosa. I tamponi in cotone Contec™ CON STIX™ offrono livelli molto bassi di residui estraibili, particelle e fibre.
Berkshire™ Open-Cell Foam Lab-Tips™ Swabs
General-purpose swab for cleaning small and confined areas
Texwipe™ CleanTip™ Swabs: Foam Series
Certified to be silicone-free
Contec™ CONSTIX™ Swabs
Polyurethane foam adhesive-bonded to a flexible nylon tip, compatible with many common solvents. Swabs are not recommended for use with ketones such as acetone or MEK.
Berkshire™ Knitted Polyester Lab-Tips Swabs
For cleaning grooved, slotted and recessed areas
Berkshire™ Open-Cell Foam Lab-Tips™ Swabs
General-purpose swab for cleaning small and confined areas.
Texwipe™ CleanTip™ Swabs: High-Sorbency Series
Certified to be silicone free
Texwipe™ Microdenier Swabs
Made of microdenier material with complete thermal bond construction that eliminates adhesive contamination
Texwipe™ CleanTip™ Swabs: Transplex™/ESD Series
ESD control without contaminating carbon or metal loading
Texwipe™ Micro CleanFoam™ ESD Swab
Use where ESD may be a concern
Texwipe™ Large Absorbond™ Sampling Swab
For cleaning validation of equipment used in manufacturing APIs, excipients, drugs, and biotech products
Texwipe™ Micro Absorbond™ ESD Swab
An excellent dissipative swab for cleaning very small grooved, slotted and recessed areas
Texwipe™ General Purpose Swabs
Designed for industrial or general-purpose applications
Texwipe™ CleanFoam Sterile Foam Swabs
Texwipe CleanFoam Sterile Foam Swabs, Length: 5 in., 132.2 mm, Width Head: 0.78 in., 19.7 mm, Handle Type: Polypropylene, Polyurethane, Recommended Applications: Safety
Contec™ CONSTIX™ Sealed Polyester Swabs
CONSTIX™ Sealed Polyester Swabs consists of 100% polyester fabric that is thermally bonded to a polypropylene handle.
Texwipe™ ESD-Safe Stick
ESD-Safe Stick with Micro-Tips is a double-ended cleaning and prepping tool designed for use at ESD-safe workstations
Texwipe™ Nylon MicroSwabs™
Texwipe’s Precision Nylon Cleanroom Swab is made from a 0.9 mm diameter tip constructed of bundled nylon filaments in a tube crimped into a 100% polypropylene handle. The filaments form a mini brush at the very tip.
Texwipe™ Sterile Polystyrene Swabs
Texwipe’s Sterile Spun Polyester Swabs are constructed using 100% USP-grade polyester, securely bonded to the wooden or polystyrene handle by a water-based adhesive.
Berkshire™ Large Closed-Cell Foam Lab-Tips™ Swabs
The LTC125 large rectangular swab is an excellent general purpose swab