Learn More
CD20 Mouse, anti-Human, R718, Clone: 2H7, BD Horizon™
Mouse Monoclonal Antibody
Marca: BD Biosciences 566988
269.64 EUR valido fino al 2025-12-31
Usa il codice promo "25339" per avere il tuo prezzo promozionale.
Descrizione
The 2H7 monoclonal antibody specifically binds to CD20, encoded by the MS4A1 (Membrane-spanning 4-domains, subfamily A, member 1) gene. CD20 is a 33-37 kDa, unglycosylated four-transmembrane phosphoprotein. CD20 is expressed on pre-B-cells, resting and activated B cells, and follicular dendritic cells, but not plasma cells. Low level CD20 expression is observed on a small subset of normal circulating T lymphocytes. The CD20 molecule is involved in the regulation of B-cell activation.

Specifica
CD20 | |
Monoclonal | |
R718 | |
100 Tests | |
Primary | |
Refrigerate Upon Arrival |
Flow Cytometry | |
2H7 | |
Mouse | |
RUO | |
Human | |
Liquid |
Suggerimenti di prodotto
Customers who viewed this item also viewed.
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.