Learn More
R&D Systems™ Human IL-23 High Sensitivity Magnetic Luminex™ Performance Assay
Magnetic bead-based multiplex assay for the Luminex™ MAGPIX™ platform
Marca: R&D Systems™ LBHS1716
Descrizione
- Magnetic format allows for easier wash steps
- Undergoes similar validation testing as Quantikine™ ELISA Kits
- Precise and Specific
- Requires a small sample volume (<50 μL)
- Run your assay in just 3-5 hours
- Mass-calibrated standards for consistent results with every new lot of material
- Enough reagents for one 96-well plate

Specifica
Magnetic Bead-based Multiplex Assay for the Luminex™ MAGPIX™ Platform | |
Human | |
Magnetic | |
17.09 - 70000pg/mL (Serum, Heparin Plasma, EDTA Plasma), 17.09 - 17500pg/mL (Cell Culture Supernates) | |
IL23A | |
Cell Culture Supernates (100μL),Serum (50μL),EDTA Plasma (50μL),Heparin Plasma (50μL) | |
Store the unopened product at 2° to 8°C. Do not use past expiration date. |
1 Kit | |
IL-23A/IL-23 P19 | |
9.52pg/mL | |
51561 | |
Serum, Plasma (EDTA, Heparin), Cell Culture Supernatant | |
|
Suggerimenti di prodotto
Customers who viewed this item also viewed.
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.